Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLCN4
Entrez Gene ID
511699
Gene Name
chloride voltage-gated channel 4
Gene Aliases
-
Chromosome Location
Chr.X: 132545378 - 132617272 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031757.1 NP_001026927.1 10 - 11 1863 65
GenBank mRNA BC151324.1 - 9 - 10 1642 65
DQ102705.1 - 10 - 11 1863 65

View Full Product Details