Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFF
Entrez Gene ID
506291
Gene Name
mitochondrial fission factor
Gene Aliases
C2H2orf33, C2orf33
Chromosome Location
Chr.2: 115486522 - 115515501 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034267.2 NP_001029439.1 6 - 7 748 89
GenBank mRNA BC102501.1 - 6 - 7 749 89
BC142371.1 - 8 - 9 1120 89

View Full Product Details