Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAGLN3
Entrez Gene ID
515562
Gene Name
transgelin 3
Gene Aliases
-
Chromosome Location
Chr.1: 56759807 - 56773635 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034499.2 NP_001029671.1 1 - 2 294 81
GenBank mRNA BC103040.1 - 1 - 2 295 81

View Full Product Details