Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DEDD
Entrez Gene ID
538428
Gene Name
death effector domain containing
Gene Aliases
-
Chromosome Location
Chr.3: 8363029 - 8374025 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034643.2 NP_001029815.1 2 - 3 80 90
GenBank mRNA BC102576.1 - 2 - 3 81 90

View Full Product Details