Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RWDD1
Entrez Gene ID
541216
Gene Name
RWD domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 34136376 - 34154203 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034726.2 NP_001029898.1 4 - 5 513 175
GenBank mRNA BC102910.1 - 4 - 5 514 175

View Full Product Details