Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPCDC
Entrez Gene ID
614957
Gene Name
phosphopantothenoylcysteine decarboxylase
Gene Aliases
MDS018
Chromosome Location
Chr.21: 33649584 - 33696731 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034773.1 NP_001029945.1 1 - 2 238 74
GenBank mRNA BC113213.1 - 1 - 2 128 74
BT021679.1 - 1 - 2 238 74

View Full Product Details