Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DFFB
Entrez Gene ID
512730
Gene Name
DNA fragmentation factor subunit beta
Gene Aliases
-
Chromosome Location
Chr.16: 49213722 - 49228162 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035109.1 NP_001030281.1 3 - 4 548 97
GenBank mRNA BC149123.1 - 4 - 5 600 97
BT021807.1 - 3 - 4 548 97

View Full Product Details