Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC42SE1
Entrez Gene ID
614042
Gene Name
CDC42 small effector 1
Gene Aliases
-
Chromosome Location
Chr.3: 19726950 - 19734579 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035112.1 NP_001030284.1 2 - 3 435 55
GenBank mRNA BC102825.1 - 2 - 3 435 55
BT021151.1 - 2 - 3 418 55

View Full Product Details