Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKG7
Entrez Gene ID
513006
Gene Name
natural killer cell granule protein 7
Gene Aliases
-
Chromosome Location
Chr.18: 57439878 - 57442652 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035279.1 NP_001030356.1 1 - 2 271 106
GenBank mRNA BC105575.1 - 1 - 2 300 106
BT021488.1 - 1 - 2 271 106

View Full Product Details