Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAR1
Entrez Gene ID
515057
Gene Name
GAR1 ribonucleoprotein
Gene Aliases
NOLA1
Chromosome Location
Chr.6: 15597104 - 15604907 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035303.1 NP_001030380.1 1 - 2 239 101
GenBank mRNA BT021549.1 - 1 - 2 239 101

View Full Product Details