Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPA33
Entrez Gene ID
518646
Gene Name
glycoprotein A33
Gene Aliases
-
Chromosome Location
Chr.3: 1675938 - 1728606 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035330.2 NP_001030407.1 3 - 4 488 61
GenBank mRNA BC102517.1 - 3 - 4 491 61

View Full Product Details