Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDLIM1
Entrez Gene ID
614675
Gene Name
PDZ and LIM domain 1
Gene Aliases
-
Chromosome Location
Chr.26: 16743890 - 16784516 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035455.1 NP_001030532.1 4 - 5 632 76
GenBank mRNA BC146213.1 - 4 - 5 653 76
BT020979.1 - 4 - 5 632 76

View Full Product Details