Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COMMD4
Entrez Gene ID
616960
Gene Name
COMM domain containing 4
Gene Aliases
-
Chromosome Location
Chr.21: 33481862 - 33487330 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040597.1 NP_001035687.1 - 105 58
GenBank mRNA BC122799.1 - 2 - 3 125 58
BT020731.1 - 2 - 3 90 58
BT020811.1 - 2 - 3 110 58
BT020814.1 - 2 - 3 96 58
BT020820.1 - 2 - 3 102 58
BT020878.1 - 2 - 3 108 58
BT020882.1 - 2 - 3 94 58
BT020897.1 - 2 - 3 97 58
BT020914.1 - - 105 58
BT020930.1 - 2 - 3 97 58
BT020940.1 - 2 - 3 103 58
BT021643.1 - 2 - 3 99 58
BT021894.1 - 2 - 3 103 58

View Full Product Details