Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERTAD3
Entrez Gene ID
767935
Gene Name
SERTA domain containing 3
Gene Aliases
-
Chromosome Location
Chr.18: 49774653 - 49779019 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077008.1 NP_001070476.1 1 - 2 66 85
GenBank mRNA BC103472.1 - 1 - 2 66 85

View Full Product Details