Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMAB
Entrez Gene ID
617636
Gene Name
metabolism of cobalamin associated B
Gene Aliases
ATR
Chromosome Location
Chr.17: 63628778 - 63639339 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079632.1 NP_001073100.1 4 - 5 380 68
GenBank mRNA BT021748.1 - 4 - 5 380 68

View Full Product Details