Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD3E
Entrez Gene ID
281054
Gene Name
CD3e molecule
Gene Aliases
-
Chromosome Location
Chr.15: 29004554 - 29015196 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174011.3 NP_776436.1 5 - 6 413 61
GenBank mRNA BC102243.1 - 5 - 6 413 61
U25687.1 - 4 - 5 325 61

View Full Product Details