Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOXL1
Entrez Gene ID
281903
Gene Name
lysyl oxidase like 1
Gene Aliases
-
Chromosome Location
Chr.21: 34663476 - 34687411 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174383.2 NP_776808.1 5 - 6 1784 71
GenBank mRNA AF421185.1 - 5 - 6 1784 71
BC126600.1 - 5 - 6 1968 71

View Full Product Details