Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOZ1
Entrez Gene ID
281939
Gene Name
myozenin 1
Gene Aliases
-
Chromosome Location
Chr.28: 29615077 - 29627523 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174397.2 NP_776822.1 3 - 4 252 79
GenBank mRNA AF359567.1 - 3 - 4 252 79
BC122689.1 - 2 - 3 124 79

View Full Product Details