Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTGIS
Entrez Gene ID
282021
Gene Name
prostaglandin I2 synthase
Gene Aliases
-
Chromosome Location
Chr.13: 77590822 - 77642755 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174444.1 NP_776869.1 9 - 10 1375 55
GenBank mRNA BC120087.1 - 9 - 10 1368 55
D30718.1 - 9 - 10 1375 55
L34208.1 - 9 - 10 1582 55

View Full Product Details