Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLA2G15
Entrez Gene ID
282271
Gene Name
phospholipase A2 group XV
Gene Aliases
ACS, LLPL, LPLA2, LYPLA3
Chromosome Location
Chr.18: 35624573 - 35638372 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174560.2 NP_776985.2 4 - 5 611 85
GenBank mRNA AY072914.1 - 4 - 5 484 85
BT021838.1 - 4 - 5 611 85

View Full Product Details