Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CKM
Entrez Gene ID
286822
Gene Name
creatine kinase, M-type
Gene Aliases
CPK-M
Chromosome Location
Chr.18: 52956486 - 52965891 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174773.4 NP_777198.2 4 - 5 553 86
GenBank mRNA AF120106.1 - 3 - 4 478 86
BC102907.1 - 4 - 5 554 86
BT020785.1 - 4 - 5 536 86
BT021173.1 - 4 - 5 539 86

View Full Product Details