Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM21
Entrez Gene ID
359715
Gene Name
tripartite motif containing 21
Gene Aliases
-
Chromosome Location
Chr.15: 50637411 - 50650199 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182655.2 NP_872596.1 5 - 6 899 70
GenBank mRNA AY312278.1 - 5 - 6 900 70
BC123700.1 - 5 - 6 902 70
BT021885.1 - 6 - 7 932 70

View Full Product Details