Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IGIP
Entrez Gene ID
404059
Gene Name
IgA inducing protein
Gene Aliases
-
Chromosome Location
Chr.7: 51314372 - 51314783 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205771.2 NP_991340.1 1 - 1 254 67
GenBank mRNA AF173827.1 - 1 - 1 254 67
BC122809.1 - 1 - 1 340 67

View Full Product Details