Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF5A
Entrez Gene ID
444990
Gene Name
eukaryotic translation initiation factor 5A
Gene Aliases
-
Chromosome Location
Chr.19: 27015088 - 27020478 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003658.2 NP_001003658.1 5 - 6 615 69
GenBank mRNA AJ577608.1 - 5 - 6 539 69
AJ577609.1 - 5 - 6 541 69
BC108135.1 - 5 - 6 615 69

View Full Product Details