Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD3D
Entrez Gene ID
281053
Gene Name
CD3d molecule
Gene Aliases
-
Chromosome Location
Chr.15: 29019453 - 29023621 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034033.2 NP_001029205.1 4 - 5 477 82
GenBank mRNA BC102857.1 - 4 - 5 478 82

View Full Product Details