Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNCA
Entrez Gene ID
282857
Gene Name
synuclein alpha
Gene Aliases
-
Chromosome Location
Chr.6: 34852799 - 35000324 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034041.2 NP_001029213.1 4 - 5 366 83
GenBank mRNA BC102402.1 - 4 - 5 368 83
BC149585.1 - 1 - 2 131 83

View Full Product Details