Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CES2
Entrez Gene ID
505928
Gene Name
carboxylesterase 2
Gene Aliases
-
Chromosome Location
Chr.18: 34638138 - 34650199 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034260.2 NP_001029432.1 8 - 9 1238 69
GenBank mRNA BC102288.1 - 8 - 9 1239 69

View Full Product Details