Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX12
Entrez Gene ID
506012
Gene Name
testis expressed 12
Gene Aliases
-
Chromosome Location
Chr.15: 22501635 - 22509202 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034263.1 NP_001029435.1 4 - 5 299 141
GenBank mRNA BC102531.1 - 4 - 5 299 141

View Full Product Details