Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHMT2
Entrez Gene ID
507197
Gene Name
serine hydroxymethyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.5: 56208140 - 56216259 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034282.1 NP_001029454.1 7 - 8 900 117
GenBank mRNA BC103242.1 - 7 - 8 900 117

View Full Product Details