Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC28B
Entrez Gene ID
507607
Gene Name
coiled-coil domain containing 28B
Gene Aliases
-
Chromosome Location
Chr.2: 121319780 - 121323914 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034305.2 NP_001029477.1 4 - 5 565 76
GenBank mRNA BC102209.1 - 4 - 5 566 76

View Full Product Details