Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLPBP
Entrez Gene ID
509643
Gene Name
pyridoxal phosphate binding protein
Gene Aliases
PROSC
Chromosome Location
Chr.27: 33031517 - 33052082 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034357.1 NP_001029529.1 2 - 3 227 81
GenBank mRNA BC102405.1 - 2 - 3 227 81

View Full Product Details