Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM45B
Entrez Gene ID
510305
Gene Name
transmembrane protein 45B
Gene Aliases
-
Chromosome Location
Chr.29: 36040192 - 36069672 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034375.2 NP_001029547.1 4 - 5 648 96
GenBank mRNA BC102148.1 - 4 - 5 649 96

View Full Product Details