Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFRP2
Entrez Gene ID
510821
Gene Name
secreted frizzled related protein 2
Gene Aliases
-
Chromosome Location
Chr.17: 3834470 - 3843043 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034393.1 NP_001029565.1 2 - 3 822 137
GenBank mRNA BC104536.1 - 2 - 3 822 137

View Full Product Details