Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRAPPC6A
Entrez Gene ID
512261
Gene Name
trafficking protein particle complex 6A
Gene Aliases
-
Chromosome Location
Chr.18: 52816715 - 52826341 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034420.2 NP_001029592.1 5 - 6 670 92
GenBank mRNA BC102525.1 - 5 - 6 671 92

View Full Product Details