Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMD9
Entrez Gene ID
513315
Gene Name
proteasome 26S subunit, non-ATPase 9
Gene Aliases
-
Chromosome Location
Chr.17: 53423947 - 53438693 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034441.2 NP_001029613.1 1 - 2 149 72
GenBank mRNA BC103247.1 - 1 - 2 152 72

View Full Product Details