Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDLIM4
Entrez Gene ID
515410
Gene Name
PDZ and LIM domain 4
Gene Aliases
-
Chromosome Location
Chr.7: 22148150 - 22162591 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034491.2 NP_001029663.1 1 - 2 138 63
GenBank mRNA BC102621.1 - 1 - 2 140 63

View Full Product Details