Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPSB3
Entrez Gene ID
515661
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
Gene Aliases
-
Chromosome Location
Chr.25: 1355611 - 1361300 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034506.1 NP_001029678.1 6 - 7 765 86
GenBank mRNA BC104512.1 - 6 - 7 765 86

View Full Product Details