Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HPX
Entrez Gene ID
534509
Gene Name
hemopexin
Gene Aliases
-
Chromosome Location
Chr.15: 46606398 - 46615970 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034612.2 NP_001029784.1 6 - 7 730 79
GenBank mRNA BC102687.1 - 6 - 7 731 79

View Full Product Details