Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASNSD1
Entrez Gene ID
539672
Gene Name
asparagine synthetase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.2: 6665580 - 6676432 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034680.2 NP_001029852.1 4 - 5 1851 62
GenBank mRNA BC103382.1 - 4 - 5 1852 62
BT026275.1 - 4 - 5 1875 62

View Full Product Details