Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTKN
Entrez Gene ID
539691
Gene Name
rhotekin
Gene Aliases
-
Chromosome Location
Chr.11: 10170661 - 10201731 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034681.1 NP_001029853.1 10 - 11 1344 94
GenBank mRNA BC102397.1 - 10 - 11 1344 94

View Full Product Details