Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSR2
Entrez Gene ID
511210
Gene Name
TSR2 ribosome maturation factor
Gene Aliases
-
Chromosome Location
Chr.X: 91960654 - 91965628 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035097.2 NP_001030269.1 1 - 2 100 91
GenBank mRNA BC102505.1 - 1 - 2 101 91

View Full Product Details