Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL45
Entrez Gene ID
511479
Gene Name
mitochondrial ribosomal protein L45
Gene Aliases
-
Chromosome Location
Chr.19: 38966103 - 38985587 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035100.2 NP_001030272.1 2 - 3 267 108
GenBank mRNA BC102136.1 - 2 - 3 270 108

View Full Product Details