Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAR1B
Entrez Gene ID
515999
Gene Name
secretion associated Ras related GTPase 1B
Gene Aliases
-
Chromosome Location
Chr.7: 46133105 - 46163331 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035315.2 NP_001030392.1 2 - 3 156 142
GenBank mRNA BC102266.1 - 2 - 3 157 142

View Full Product Details