Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SC5D
Entrez Gene ID
525154
Gene Name
sterol-C5-desaturase
Gene Aliases
SC5DL
Chromosome Location
Chr.15: 31876089 - 31922383 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035356.1 NP_001030433.1 2 - 3 290 149
GenBank mRNA BC103338.1 - 2 - 3 290 149

View Full Product Details