Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CMAS
Entrez Gene ID
100336710
Gene Name
cytidine monophosphate N-acetylneuraminic acid synthetase
Gene Aliases
-
Chromosome Location
Chr.5: 88154147 - 88169865 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035398.1 NP_001030475.1 1 - 2 285 132
GenBank mRNA BC102786.1 - 1 - 2 285 132

View Full Product Details