Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
QPRT
Entrez Gene ID
614254
Gene Name
quinolinate phosphoribosyltransferase
Gene Aliases
-
Chromosome Location
Chr.25: 26491479 - 26506183 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035446.2 NP_001030523.1 1 - 2 59 62
GenBank mRNA BC102550.1 - 1 - 2 60 62

View Full Product Details