Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPL3L
Entrez Gene ID
618440
Gene Name
ribosomal protein L3 like
Gene Aliases
-
Chromosome Location
Chr.25: 1507603 - 1514414 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035501.1 NP_001030578.1 7 - 8 969 68
GenBank mRNA BC103272.1 - 7 - 8 969 68

View Full Product Details