Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL3L
Entrez Gene ID
618440
Gene Name
ribosomal protein L3 like
Gene Aliases
-
Chromosome Location
Chr.25: 1507603 - 1514414 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035501.1 NP_001030578.1 9 - 10 1192 66
GenBank mRNA BC103272.1 - 9 - 10 1192 66

View Full Product Details