Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RANGRF
Entrez Gene ID
512506
Gene Name
RAN guanine nucleotide release factor
Gene Aliases
RANGNRF
Chromosome Location
Chr.19: 27903878 - 27905325 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037460.2 NP_001032537.1 2 - 3 313 91
GenBank mRNA BC108151.1 - 2 - 3 314 91

View Full Product Details