Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2H2orf76
Entrez Gene ID
613457
Gene Name
chromosome 2 C2orf76 homolog
Gene Aliases
-
Chromosome Location
Chr.2: 71107880 - 71214464 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037618.2 NP_001032707.1 6 - 7 473 111
GenBank mRNA BC109869.1 - 6 - 7 474 111

View Full Product Details